missing translation for 'onlineSavingsMsg'
Learn More

Excelta™ Medical-Grade Scissors

Catalog No. 17456007
Change view
Click to view available options
Blade Style:
Curved
Curved, Fine
Curved, Very Fine
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Blade Style
17456007 Curved
17456006 Curved, Very Fine
17456005 Curved, Fine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17456007 Supplier Excelta™ Supplier No. XLT363

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Made of 316L stainless steel

  • Curved blades

Specifications

Blade Style Curved
Grade Medical
Length (English) 3.25 in.
Length (Metric) 8.25 cm
For Use With (Application) Surgical, Precision
Tip Style Round