missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Mdivi 1

Catalog No. 398210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
398210 10 mg
398250 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 398210 Supplier Tocris Bioscience™ Supplier No. 3982/10*
Only null left

Selective dynamin inhibitor; attenuates mitochondrial division and apoptosis

Mdivi 1 is a selective inhibitor of dynamin GTPase (IC50 = 1 - 10 μM); attenuates Dnm1- and Drp1-mediated mitochondrial division. Mdivi 1 inhibits mitochondrial outer membrane permeabilization (MOMP) and attenuates apoptosis. In cancer cell lines, Mdivi 1 reduces cell proliferation; in an animal model of ischemic retina Mdivi 1 treatment blocks apoptosis. In a cell model, Mdivi 1 protects against mutant huntington-induced damage and mitochondrial dysfunction. Mdivi 1 also inhibits autophagy and protects neurons from excitotoxicity.

Chemical Identifiers

CAS 338967-87-6
Molecular Formula C15H10Cl2N2O2S

Specifications

Quantity 10 mg
Formula Weight 353.22
Purity >98%
Molecular Weight (g/mol) 353.217
Chemical Name or Material Dynamin Inhibitors
Recommended Storage Store at -20°C