missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ mCherry Monoclonal Antibody (16D7)

Description
mCherry Monoclonal Antibody for Flow, ICC/IF, IHC (P), IP, Western Blot
MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.
Spécifications
Spécifications
| Antigène | mCherry |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 16D7 |
| Concentration | 2 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 0.09% sodium azide |
| Alias du gène | mCherry |
| Espèces hôtes | Rat |
| Immunogène | full-length protein mCherry. |
| Afficher plus de résultats |