missing translation for 'onlineSavingsMsg'
Learn More

MBTH Indicator, 0.05% (w/v), Ricca Chemical

Catalog No. 462232
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
462232 1 L Amber Glass Bottle
46224 120 mL Amber Glass Bottle
462216 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 462232 Supplier Ricca Chemical Company Supplier No. 462232

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

APHA/ASTM for Cyanide Testing

Specifications

CAS 149022-15-1, 7732-18-5
CAS Min % 98.0, 0.05
CAS Max % 100.0, 0.05
Molecular Formula C8H10ClN3S
InChI Key OEZPVSPULCMUQB-DQMXGCRQSA-N
SMILES CN1C2=CC=CC=C2SC1=NN.Cl
IUPAC Name (Z)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrochloride
Molecular Weight (g/mol) 215.699
PubChem CID 9575839
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
Color Colorless
Quantity 1 L
Chemical Name or Material MBTH Indicator
Show More Show Less