Learn More
Promega Maxwell 16 High Strength LEV Magnetic Rod and Plunger Bar Adaptor, 1 each DFS Item
Accessories for the Maxwell(R) 16 and Maxwell(R) 16 Forensic Instruments in either SEV or LEV Configuration
Supplier:  Promega SP1070
Catalog No. PRSP1070
Only null left
Explore available promotions
Description
Description
  • Accessories for the Maxwell(R) 16 and Maxwell(R) 16 Forensic Instruments in either SEV or LEV Configuration
  • Standard Elution Volume (SEV) is 200−400μl
  • Low Elution Volume (LEV) is 30−100μl
  • Plungers and Elution Tubes for the Maxwell(R) 16 Instruments only (AS2000 and AS3000)
Specifications
Specifications
1 Ea.
Product Suggestions
Product Suggestions