Learn More
Electron Microscopy Sciences NUNC MaxiSorp™ 96 Well Plates (Polystyrene) Clear
Numéro de catalogue. 5031451
Encompass
NUNC MaxiSorp plates have been the certified ELISA plate industry standard for years. This same surface now sets a new standard for protein multiplexing. Certified for absorption of IgG; CV of <5% between wells; all results are within 10% from the mean and background of all well within 0.005 absorbance units from the mean Plate: Clear Polystyrene for Protein & antibody multiplexing, chemiluminescence.
Numéro de catalogue. 5031451 Fournisseur Electron Microscopy Sciences Code fournisseur 63494CL
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
NUNC MaxiSorp plates have been the certified ELISA plate industry standard for years. This same surface now sets a new standard for protein multiplexing. Certified for absorption of IgG; CV of <5% between wells; all results are within 10% from the mean and background of all well within 0.005 absorbance units from the mean Plate: Clear Polystyrene for Protein & antibody multiplexing, chemiluminescence.