missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Matrix oil, 75 centistoke hydrocarbon oil, Thermo Scientific Chemicals
$410.86 - $410.86
Chemical Identifiers
| CAS | 8042-47-5 |
|---|---|
| Molecular Formula | C16H10N2Na2O7S2 |
| Molecular Weight (g/mol) | 452.363 |
| MDL Number | MFCD00131611 |
| InChI Key | AEOVEGJBKQQFOP-DDVLFWKVSA-L |
| PubChem CID | 9566064 |
| IUPAC Name | disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate |
| SMILES | C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA14621EQC
|
Thermo Scientific Chemicals
014621EQC |
3.8 L |
Each for $410.86
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 8042-47-5 | |
| 452.363 | |
| AEOVEGJBKQQFOP-DDVLFWKVSA-L | |
| disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate |
| C16H10N2Na2O7S2 | |
| MFCD00131611 | |
| 9566064 | |
| C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] |
Specifications
| Plastic bottle | |
| >110°C (230°F) | |
| C16H10N2Na2O7S2 | |
| AEOVEGJBKQQFOP-DDVLFWKVSA-L | |
| disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate | |
| 3.8 L | |
| Matrix oil |
| 8042-47-5 | |
| Oily Liquid | |
| MFCD00131611 | |
| C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+] | |
| 452.363 | |
| 9566064 |
Safety and Handling
P301+P310-P331-P501c
H304
EINECSNumber : 232-455-7
RTECSNumber : PY8047000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only