missing translation for 'onlineSavingsMsg'
Learn More

Matrix oil, 75 centistoke hydrocarbon oil, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 014621EQC

 View more versions of this product

Catalog No. AA14621EQC

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Matrix oil
8042-47-5
Oily Liquid
MFCD00131611
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
452.363
9566064
Plastic bottle
>110°C (230°F)
C16H10N2Na2O7S2
AEOVEGJBKQQFOP-DDVLFWKVSA-L
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
3.8 L

Chemical Identifiers

8042-47-5
452.363
AEOVEGJBKQQFOP-DDVLFWKVSA-L
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
C16H10N2Na2O7S2
MFCD00131611
9566064
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P301+P310-P331-P501c

H304

EINECSNumber : 232-455-7

RTECSNumber : PY8047000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only