missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Matrix Modifier, 40% (w/v) Ammonium Phosphate in Water, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Matrix Modifier |
| Packaging | Natural Poly Bottle |
| Name Note | 40% (w/v) Ammonium Phosphate |
| CAS | 7783-28-0, 7732-18-5 |
| CAS Min % | 59.0, 39.0 |
| CAS Max % | 61.0, 41.0 |