missing translation for 'onlineSavingsMsg'
Learn More

StatLab MasterTech Hydroquinone Capsules

Catalog No. 22035368
Change view
Click to view available options
Quantity:
25/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22035368 25/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22035368 Supplier StatLab Supplier No. AHH0374P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or if inhaled

A StatLab MasterTech Special Stain Kit Component

  • Hydroquinone Capsules are a component for the following MasterTech Stain Kits Steiner Stain Kit Dieterles Stain Kit and Modified Genta Triple Stain Kit

Specifications

Physical Form Solution
CAS 123-31-9
Color White
Quantity 25/Pk.
Grade Laboratory