missing translation for 'onlineSavingsMsg'
Learn More

StatLab MasterTech 2% Oxalic Acid

Catalog No. 21403475
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21403475 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21403475 Supplier StatLab Supplier No. AHO1126

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or if inhaled

For Bleaching and Decolorization of Tissue Samples.

  • Used to remove residual potassium permanganate from tissue sections
  • It ensures the tissue is properly prepared for subsequent steps in staining reticulin fibers, specifically after oxidation and before silver impregnation
  • Solution can be used for removing calcium from tissue samples
  • A StatLab MasterTech Special Stain Kit component

Specifications

Physical Form Solution
CAS 6153-56-6
Color Clear Colorless
Quantity 100 mL
Grade Laboratory