missing translation for 'onlineSavingsMsg'
Learn More

StatLab MasterTech 1% Sodium Bisulfite

Catalog No. 21010304
Change view
Click to view available options
Color:
Colorless
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21010304 Colorless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21010304 Supplier StatLab Supplier No. KC0943

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or if inhaled

Utilized to qualitatively demonstrate fungi and Pneumocystis carinii.

  • A component for the MasterTech GMS Stain Kit
  • A MasterTech Special Stain Kit Component

Specifications

Physical Form Solution
CAS 7631-90-5
Color Colorless
Quantity 1 L
Grade Laboratory