missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mast Cell Chymase Recombinant Rabbit Monoclonal Antibody (23GB5095)
GREENER_CHOICE

Numéro de catalogue. MA552788
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MA552788 100 μL
MA552789 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MA552788 Fournisseur Invitrogen™ Code fournisseur MA552788
Il en reste null

Rabbit Recombinant Monoclonal Antibody

Mast Cell Chymase Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene product is a chymotryptic serine proteinase that belongs to the peptidase family S1. It is expressed in mast cells and thought to function in the degradation of the extracellular matrix, the regulation of submucosal gland secretion, and the generation of vasoactive peptides. In the heart and blood vessels, this protein, rather than angiotensin converting enzyme, is largely responsible for converting angiotensin I to the vasoactive peptide angiotensin II. Angiotensin II has been implicated in blood pressure control and in the pathogenesis of hypertension, cardiac hypertrophy, and heart failure. Thus, this gene product is a target for cardiovascular disease therapies. This gene maps to 14q11. 2 in a cluster of genes encoding other proteases.

Spécifications

Antigen Mast Cell Chymase
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5095
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CMA1
Gene Accession No. P21844
Gene Alias alpha-chymase; Chymase; chymase 1; chymase 1 preproprotein transcript E; chymase 1 preproprotein transcript I; chymase 1, mast cell; chymase, heart; chymase, mast cell; CMA1; CYH; CYM; mast cell; Mast cell chymase 1; mast cell protease 3; Mast cell protease 5; mast cell protease I; Mast cell protease III; MCP3P; Mcp5; Mcp-5; Mcpt3; Mcpt5; MCT1; mMCP-5; rMCP-3; rMCP-5; rMCP-III
Gene Symbols CMA1
Host Species Rabbit
Immunogen A synthesized peptide derived from human Mast Cell Chymase (204-247AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17228
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats