missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MARCO Monoclonal Antibody (PLK-1)
GREENER_CHOICE

Numéro de catalogue. MA140315
Modifier l'affichage
Click to view available options
Quantité:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
MA140315 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. MA140315 Fournisseur Invitrogen™ Code fournisseur MA140315
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Antibody

Endotoxin: 0.40 EU/mg Application Notes: IHC (frozen) - tissue sections can be fixed in acetone or 2% paraformaldehyde. Western blot - use non-reducing conditions. Flow - this antibody weakly stains alveolar macrophages by recognizing the extracellular domain of MARCO, with transfected COS cells used as a positive control. Immunoprecipitation - will immunoprecipitate MARCO as a 60 kDa and 50 kDa protein from transfected COS lysates. Functional Assays - blocks human alveolar macrophages by recognizing the extracellular doamin of MARCO. Recommended positive control: Human alveolar macrophages; Negative control: All other human cells.

MARCO, also known as SR-A6 or SCARA2, is a macrophage surface receptor and a member of the class A scavenger receptor family. As a part of the innate antimicrobial immune system, MARCO can bind both Gram-negative and Gram-positive bacteria via an extracellular, C-terminal, scavenger receptor cysteine-rich (SRCR) domain. In addition to short cytoplasmic and transmembrane domains, there is an extracellular spacer domain and a long, extracellular collagenous domain. The protein may form a trimeric molecule by the association of the collagenous domains of three identical polypeptide chains. Due to its expression in tumor infiltrating macrophages, and its role in inhibiting immune responses within tumor environment, MARCO has been indicated as a potential target for cancer immunotherapy.

Spécifications

Antigène MARCO
Applications Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone PLK-1
Concentration 0.1 mg/mL
Conjugué Unconjugated
Formule PBS with 0.1% BSA and no preservative
Gène Marco
Numéro d’acquisition du gène Q9UEW3
Alias du gène AI323439; Ly112; macrophage receptor MARCO; Macrophage receptor with collagenous structure; MARCO; SCARA2; Scavenger receptor class A member 2; scavenger receptor class A, member 2; SR-A6
Symboles de gène Marco
Espèces hôtes Mouse
Immunogène Human alveolar macrophages (recognizes domain V between residues 420 and 431).
Méthode de purification Protein G
Quantité 100 μg
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 505632, 8685
Espèces cibles Human, Bovine
Contenu et stockage 4°C
Type de produit Antibody
Forme Liquid
Isotype IgG3
Afficher plus de résultats Afficher moins de résultats