missing translation for 'onlineSavingsMsg'
Learn More

Mannose triflate, 98%

Numéro de catalogue. p-7077880
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC461341000 100 mg
AC461340010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC461341000 Fournisseur Thermo Scientific Chemicals Code fournisseur 461341000
Il en reste null

CAS: 92051-23-5 | C15H19F3O12S | 480.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 92051-23-5
Molecular Formula C15H19F3O12S
Molecular Weight (g/mol) 480.36
InChI Key OIBDVHSTOUGZTJ-PEBLQZBPSA-N
Synonym tatm, mannose triflate, 1,3,4,6-tetra-o-acetyl-2-o-trifluoromethanesulfonyl-beta-d-mannopyranose, 1,3,4,6-tetra-o-acetyl-2-o-trifluoromethanesulfonyl-b-d-mannopyranose, unii-0ki8ffq6tt, 0ki8ffq6tt, beta-d-mannopyranose 1,3,4,6-tetra-o-acetate 2-o-trifluoromethanesulfonate, 2r,3r,4s,5s,6s-3,4,6-tris acetyloxy-5-trifluoromethanesulfonyloxy oxan-2-yl methyl acetate, mannose trilfate
PubChem CID 11113597
IUPAC Name [(2R,3R,4S,5S,6S)-3,4,6-triacetyloxy-5-(trifluoromethylsulfonyloxy)oxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OS(=O)(=O)C(F)(F)F)OC(=O)C)OC(=O)C

Spécifications

Melting Point 118.0°C to 122.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Quantity 100 mg
Solubility Information Clear colorless to slightly yellow
Formula Weight 480.36
Percent Purity 98%
Chemical Name or Material Mannose triflate

RUO – Research Use Only