missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mannitol, Reagent, ACS, Spectrum™ Chemical
M1120, 69-65-8, C6H14O6
$336.70 - $12202.48
Chemical Identifiers
| CAS | 69-65-8 |
|---|---|
| Molecular Formula | C6H14O6 |
| Molecular Weight (g/mol) | 182.17 |
| MDL Number | MFCD00064287 |
| InChI Key | FBPFZTCFMRRESA-UHFFFAOYNA-N |
| IUPAC Name | hexane-1,2,3,4,5,6-hexol |
| SMILES | OCC(O)C(O)C(O)C(O)CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18604229
|
Spectrum Chemical Mfg Cor
M1120125GM |
125 g | Poly Bottle |
Each for $336.70
|
|
||||
|
18604230
|
Spectrum Chemical Mfg Cor
M1120500GM |
500 g | Poly Bottle |
Each for $626.42
|
|
||||
|
18604232
|
Spectrum Chemical Mfg Cor
M112025KG |
2.5 kg | Poly Pail |
Each for $799.70
|
|
||||
|
18604231
|
Spectrum Chemical Mfg Cor
M1120500GMCS6 |
6 x 500 g | Poly Bottle |
Case of 6 Each for $2,943.58
|
|
||||
|
18604233
|
Spectrum Chemical Mfg Cor
M112025KGCS4 |
4 x 2.5 kg | Poly Pail |
Case of 4 Each for $8,067.61
|
|
||||
|
18604234
|
Spectrum Chemical Mfg Cor
M112012KG |
12 kg | Poly Pail |
Each for $7,543.52
|
|
||||
|
18604235
|
Spectrum Chemical Mfg Cor
M112050KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Mannitol, Reagent, ACS , otherwise known as Mannite, is classified as a sugar alcohol and is most commonly observed in it′s white crystalline solid form. It has several uses, ranging from inducing osmotic stress in plants to being used to produce tablets of medicine. It can also be added to boric acid to increase the strength of the acid in order to allow for a better degree of accuracy in volumetric analysis of the acid. As an ACS grade Reagent, Spectrum Chemical manufactured mannitol is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.Chemical Identifiers
| 69-65-8 | |
| 182.17 | |
| FBPFZTCFMRRESA-UHFFFAOYNA-N | |
| OCC(O)C(O)C(O)C(O)CO |
| C6H14O6 | |
| MFCD00064287 | |
| hexane-1,2,3,4,5,6-hexol |
Specifications
| 69-65-8 | |
| 0.05% | |
| C6H14O6 | |
| 125 g | |
| FBPFZTCFMRRESA-UHFFFAOYNA-N | |
| hexane-1,2,3,4,5,6-hexol | |
| ACS |
| 100% | |
| Poly Bottle | |
| MFCD00064287 | |
| +23.3° to +24.3° | |
| OCC(O)C(O)C(O)C(O)CO | |
| 182.17 |