missing translation for 'onlineSavingsMsg'
Learn More

Mannitol, Reagent, ACS, Spectrum™ Chemical

M1120, 69-65-8, C6H14O6

Supplier:  Spectrum Chemical Mfg Cor M112012KG

 View more versions of this product

Catalog No. 18604234

Only null left

Description

Description

Spectrum™ Chemical Mannitol, Reagent, ACS , otherwise known as Mannite, is classified as a sugar alcohol and is most commonly observed in it′s white crystalline solid form. It has several uses, ranging from inducing osmotic stress in plants to being used to produce tablets of medicine. It can also be added to boric acid to increase the strength of the acid in order to allow for a better degree of accuracy in volumetric analysis of the acid. As an ACS grade Reagent, Spectrum Chemical manufactured mannitol is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Specifications

Specifications

69-65-8
0.05%
C6H14O6
12 kg
FBPFZTCFMRRESA-UHFFFAOYNA-N
hexane-1,2,3,4,5,6-hexol
ACS
100%
Poly Pail
MFCD00064287
+23.3° to +24.3°
OCC(O)C(O)C(O)C(O)CO
182.17

Chemical Identifiers

69-65-8
182.17
FBPFZTCFMRRESA-UHFFFAOYNA-N
OCC(O)C(O)C(O)C(O)CO
C6H14O6
MFCD00064287
hexane-1,2,3,4,5,6-hexol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions