missing translation for 'onlineSavingsMsg'
Learn More

Mannitol, Reagent, ACS, Spectrum™ Chemical

Catalog No. 18604232
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604232 2.5 kg Poly Pail
18604229 125 g Poly Bottle
18604230 500 g Poly Bottle
18604231 6 x 500 g Poly Bottle
18604233 4 x 2.5 kg Poly Pail
18604234 12 kg Poly Pail
18604235 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18604232 Supplier Spectrum Chemical Mfg Cor Supplier No. M11202.5KG
Only null left

M1120, 69-65-8, C6H14O6

Spectrum™ Chemical Mannitol, Reagent, ACS , otherwise known as Mannite, is classified as a sugar alcohol and is most commonly observed in it′s white crystalline solid form. It has several uses, ranging from inducing osmotic stress in plants to being used to produce tablets of medicine. It can also be added to boric acid to increase the strength of the acid in order to allow for a better degree of accuracy in volumetric analysis of the acid. As an ACS grade Reagent, Spectrum Chemical manufactured mannitol is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

CAS Max % 100%
Loss on Drying 0.05%
Packaging Poly Pail
Quantity 2.5 kg
Specific Rotation +23.3° to +24.3°
Grade ACS