missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mannitol, Powder, USP, 97-102%, Spectrum™ Chemical
MA165, 69-65-8, C6H14O6
$313.51 - $11863.83
Chemical Identifiers
| CAS | 69-65-8 |
|---|---|
| Molecular Formula | C6H14O6 |
| Molecular Weight (g/mol) | 182.17 |
| MDL Number | MFCD00064287 |
| InChI Key | FBPFZTCFMRRESA-UHFFFAOYNA-N |
| IUPAC Name | hexane-1,2,3,4,5,6-hexol |
| SMILES | OCC(O)C(O)C(O)C(O)CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18604224
|
Spectrum Chemical Mfg Cor
MA165125GM |
125 g | Poly Bottle |
Each for $313.51
|
|
||||
|
18604225
|
Spectrum Chemical Mfg Cor
MA165500GM |
500 g | Poly Bottle |
Each for $1,056.06
|
|
||||
|
18604226
|
Spectrum Chemical Mfg Cor
MA16525KG |
2.5 kg | Poly Pail |
Each for $2,201.23
|
|
||||
|
18604227
|
Spectrum Chemical Mfg Cor
MA16512KG |
12 kg | Poly Pail |
Each for $6,793.00
|
|
||||
|
18604228
|
Spectrum Chemical Mfg Cor
MA16550KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Mannitol, Powder, USP is a diuretic used to increase urine production.All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 69-65-8 | |
| 182.17 | |
| FBPFZTCFMRRESA-UHFFFAOYNA-N | |
| OCC(O)C(O)C(O)C(O)CO |
| C6H14O6 | |
| MFCD00064287 | |
| hexane-1,2,3,4,5,6-hexol |
Specifications
| 69-65-8 | |
| 0.5% | |
| Poly Bottle | |
| MFCD00064287 | |
| FBPFZTCFMRRESA-UHFFFAOYNA-N | |
| hexane-1,2,3,4,5,6-hexol | |
| 97 to 102% |
| 100% | |
| 165-170°C | |
| C6H14O6 | |
| 125 g | |
| OCC(O)C(O)C(O)C(O)CO | |
| 182.17 | |
| USP |