missing translation for 'onlineSavingsMsg'
Learn More

Mannitol, Powder, USP, 97-102%, Spectrum™ Chemical

Catalog No. 18604228
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604228 50 kg Poly Pail
18604224 125 g Poly Pail
18604225 500 g Poly Pail
18604226 2.5 kg Poly Pail
18604227 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604228 Supplier Spectrum Chemical Mfg Cor Supplier No. MA16550KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MA165, 69-65-8, C6H14O6

Spectrum™ Chemical Mannitol, Powder, USP is a diuretic used to increase urine production.All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

CAS Max % 100%
Loss on Drying 0.5%
Melting Range 165-170°C
Packaging Poly Pail
Quantity 50 kg
Percent Purity 97 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Mannitol, Powder