missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mannitol, Powder, USP, 97-102%, Spectrum™ Chemical
MA165, 69-65-8, C6H14O6
Supplier: Spectrum Chemical Mfg Cor MA16512KG
Description
Spectrum™ Chemical Mannitol, Powder, USP is a diuretic used to increase urine production.All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 69-65-8 | |
| 0.5% | |
| Poly Pail | |
| MFCD00064287 | |
| FBPFZTCFMRRESA-UHFFFAOYNA-N | |
| hexane-1,2,3,4,5,6-hexol | |
| 97 to 102% |
| 100% | |
| 165-170°C | |
| C6H14O6 | |
| 12 kg | |
| OCC(O)C(O)C(O)C(O)CO | |
| 182.17 | |
| USP |
Chemical Identifiers
| 69-65-8 | |
| 182.17 | |
| FBPFZTCFMRRESA-UHFFFAOYNA-N | |
| OCC(O)C(O)C(O)C(O)CO |
| C6H14O6 | |
| MFCD00064287 | |
| hexane-1,2,3,4,5,6-hexol |