missing translation for 'onlineSavingsMsg'
Learn More

Mannitol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015265
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015265 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015265 Supplier Merck Emd Millipore Supplier No. PHR10071G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mannitol is an osmotic diuretic with a variety of clinical applications, one of which is the reduction of intracranial pressure. It is also found to be protective against ischemic and nephrotoxic acute renal failure.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 69-65-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C6H14O6
MDL Number MFCD00064287
Melting Point 167°C to 170°C
Synonym D-Mannitol; Mannite
Type Certified Reference Material
Quantity 1 g
Formula Weight 182.17
Grade Certified Reference Material
Show More Show Less