missing translation for 'onlineSavingsMsg'
Learn More

Mannitol ACS AR Powder, Macron Fine Chemicals™

Catalog No. MK6209212
Change view
Click to view available options
STAKMOR; 2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK6209212 STAKMOR; 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK6209212 Supplier Avantor Supplier No. 620905

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 69-65-8
Packaging STAKMOR
Molecular Formula C6H14O6
MDL Number MFCD00064287
Quantity 2.5kg
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
SMILES OCC(O)C(O)C(O)C(O)CO
IUPAC Name hexane-1,2,3,4,5,6-hexol
Molecular Weight (g/mol) 182.17
PubChem CID 6251
ChEBI CHEBI:16899
Formula Weight 182.17
Grade ACS
Chemical Name or Material Mannitol
Show More Show Less