missing translation for 'onlineSavingsMsg'
Learn More

Manganese Sulfate Monohydrate, Purified, Reagents

Catalog No. 01337181
Change view
Click to view available options
Quantity:
500 g
3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337181 3 kg
01337180 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337181 Supplier Labchem Inc Supplier No. C22430003A5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Manganese Sulfate Monohydrate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym Manganous Sulfate Monohydrate
IUPAC Name manganese(2+) hydrate sulfate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Boiling Point 850°C
Color Pink
Physical Form Solid
Quantity 3 kg
Percent Purity 100%
Grade Pure
Packaging Poly Bottle
Appearance Pink
Specific Gravity 2.95
Chemical Name or Material Manganese Sulfate Monohydrate, Purified