missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese Sulfate Monohydrate, ACS Reagent (Manganous Sulfate), Reagents
Specifications
Specifications
| Boiling Point | 850°C |
| Melting Point | 700°C |
| Color | Pink |
| Physical Form | Solid |
| Quantity | 100 g |
| Percent Purity | ≥98% |
| Grade | ACS |
| Packaging | Glass Bottle |
| Specific Gravity | 2.95 |
| Chemical Name or Material | Manganese Sulfate Monohydrate, ACS Reagent (Manganous Sulfate) |