missing translation for 'onlineSavingsMsg'
Learn More

Manganese(IV) oxide, 99.9% (metals basis)

Catalog No. AA4225030
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4225030 250 g
AA42250A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4225030 Supplier Thermo Scientific Chemicals Supplier No. 04225030

CAS: 1313-13-9 | MnO2 | 86.94 g/mol

It is used as an oxidizing agent in organic synthesis such as oxidation of allylic/benzylic alcohols, as a textile dye, as a reducing agent, and as a component of dry cell batteries such as zinc-carbon battery and alkaline battery. It is also used in making pigments for glasses and ceramics, and drier for paints. Further it is used in the manufacture of manganese steel and several manganese derivatives including potassium permanganate, a powerful versatile oxidant. Catalytic hydration of nitriles to amides by flowing through manganese dioxide has been reported. Manganese dioxide supported on inorganic oxide can be used for oxidation of methylamine through CWAO (Catalytic Wet Air Oxidation). It has high potential as highly efficient and robust material for water oxidation reaction (WORs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1313-13-9
Molecular Formula MnO2
Molecular Weight (g/mol) 86.94
MDL Number MFCD00003463
InChI Key NUJOXMJBOLGQSY-UHFFFAOYSA-N
Synonym manganese dioxide, manganese iv oxide, manganese peroxide, manganese superoxide, manganese black, manganese binoxide, manganese oxide, mangandioxid, black manganese oxide, braunstein
PubChem CID 14801
IUPAC Name dioxomanganese
SMILES O=[Mn]=O

Specifications

Melting Point 535°C (decomposition)
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.9% (metals basis)
Merck Index 14,5730
Solubility Information Insoluble in water.
Formula Weight 86.94
Percent Purity 99.9%
Density 5.026 g/mL
Assay (metals basis)
Chemical Name or Material Manganese(IV) oxide
Show More Show Less
Hazard Category H302+H332-H373
Hazard Statement GHS H Statement
H302-H332
Precautionary Statement P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c
EINECSNumber 215-202-6
RTECSNumber OP0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only