missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfate monohydrate, ACS, 98.0-101.0%

Catalog No. AA3334136
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3334136 500 g
AA33341A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3334136 Supplier Thermo Scientific Chemicals Supplier No. 03334136

CAS: 10034-96-5 | H2MnO5S | 169.01 g/mol

Manganese(II) sulfate monohydrate is used as a colorant in dyes, fertilizers, animal feeds and red glazes on porcelain. Further, it is used in paints, ceramics, nutrient and dietary supplement. It is involved in the preparation of manganese dioxide. In addition, it serves as a precursor to manganese metal and other manganese compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+);sulfate;hydrate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Color Pink
Physical Form Powder
Quantity 500 g
Assay Percent Range 98.0-101.0%
Solubility Information Soluble in water and alcohol. Insoluble in ether.
Formula Weight 169.02 (151.00 Anhydrous)
Grade ACS
Appearance Pink powder
Density 2.95
Chemical Name or Material Manganese(II) sulfate monohydrate
Hazard Category H318-H373
Precautionary Statement P260-P280-P305+P351+P338-P310-P314-P501c
DOTInformation DOT Class: 9; Packing Group: III
EINECSNumber 232-089-9
RTECSNumber OP0893500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only