missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) nitrate, 50% w/w aq. soln.

Catalog No. AA33340A3
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33340A3 2 kg
AA3334036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA33340A3 Supplier Thermo Scientific Chemicals Supplier No. 033340A3
Only null left

CAS: 10377-66-9 | MnN2O6 | 178.946 g/mol

Manganese(II) nitrate is used as a precursor to prepare the oxides of manganese. It is involved in the preparation of electronic components. Further, it is utilized for the phosphorous treatment of metal surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10377-66-9
Molecular Formula MnN2O6
Molecular Weight (g/mol) 178.946
MDL Number MFCD00011112
InChI Key MIVBAHRSNUNMPP-UHFFFAOYSA-N
Synonym manganese nitrate, manganous nitrate, manganese dinitrate, manganous dinitrate, manganese ii nitrate, manganese 2+ nitrate, nitric acid, manganese 2+ salt, manganese ii nitrate, anhydrous, manganesenitrate
PubChem CID 61511
IUPAC Name manganese(2+);dinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Mn+2]

Specifications

Color Pink
Physical Form Liquid
Quantity 2 kg
Solubility Information Miscible with water.
Formula Weight 178.95
Appearance Pink liquid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Chemical Name or Material Manganese(II) nitrate, 50% w/w aq. Soln
Hazard Category H271-H302-H314-H373
Precautionary Statement P210-P220-P221-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation DOT Class: 8; Packaging Group: II
EINECSNumber 233-828-8
RTECSNumber QU9780000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only