missing translation for 'onlineSavingsMsg'
Learn More

Manganese ICP Standard, 1000 ppm Mn in 3% HNO3, Ricca Chemical

Catalog No. p-3251891
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PMN1KN100 100 mL Natural LDPE Bottle
PMN1KN500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PMN1KN100 Supplier Ricca Chemical Company Supplier No. PMN1KN100
Only null left

Chemical Identifiers

CAS 6156-78-1 , 7697-37-2 , 7732-18-5
Molecular Formula C4H14MnO8
Molecular Weight (g/mol) 245.09
MDL Number MFCD00062552
InChI Key CESXSDZNZGSWSP-UHFFFAOYSA-L
PubChem CID 93021
IUPAC Name manganese(2+) diacetate tetrahydrate
SMILES O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Mn
CAS Min % 93.0, 4.21, 0.42
CAS Max % 97.0, 4.38, 0.43
Equivalence 1 mL = 1 mg Mn
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Chemical Name or Material Manganese ICP Standard
Show More Show Less