missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese ICP Standard, 10,000 ppm Mn in 5% HNO3, Ricca Chemical
$1148.00 - $1148.00
Chemical Identifiers
| CAS | 6156-78-1 |
|---|---|
| Molecular Formula | C4H14MnO8 |
| Molecular Weight (g/mol) | 245.09 |
| MDL Number | MFCD00062552 |
| InChI Key | CESXSDZNZGSWSP-UHFFFAOYSA-L |
| PubChem CID | 93021 |
| IUPAC Name | manganese(2+) diacetate tetrahydrate |
| SMILES | O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PMN10KN-500
|
Ricca Chemical Company
PMN10KN500 |
500 mL | Natural Poly Bottle |
Each for $1,148.00
|
|
||||
Chemical Identifiers
| 6156-78-1 | |
| 245.09 | |
| CESXSDZNZGSWSP-UHFFFAOYSA-L | |
| manganese(2+) diacetate tetrahydrate |
| C4H14MnO8 | |
| MFCD00062552 | |
| 93021 | |
| O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O |
Specifications
| Natural Poly Bottle | |
| 10,000ppm Mn | |
| 91.0,6.84,4.07 | |
| Liquid | |
| MFCD00062552 | |
| < 2 | |
| CESXSDZNZGSWSP-UHFFFAOYSA-L | |
| manganese(2+) diacetate tetrahydrate | |
| 500 mL | |
| Manganese ICP Standard |