missing translation for 'onlineSavingsMsg'
Learn More

Manganese ICP Standard, 10,000 ppm Mn in 5% HNO3, Ricca Chemical

Catalog No. PMN10KN500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PMN10KN500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PMN10KN500 Supplier Ricca Chemical Company Supplier No. PMN10KN500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6156-78-1 , 7697-37-2 , 7732-18-5
Molecular Formula C4H14MnO8
Molecular Weight (g/mol) 245.09
MDL Number MFCD00062552
InChI Key CESXSDZNZGSWSP-UHFFFAOYSA-L
PubChem CID 93021
IUPAC Name manganese(2+) diacetate tetrahydrate
SMILES O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 10,000ppm Mn
CAS Min % 88.0, 6.58, 3.91
CAS Max % 91.0, 6.84, 4.07
Equivalence 1 mL = 10 mg Mn
Physical Form Liquid
Color Pink
pH < 2
Type Standard
Quantity 500 mL
Chemical Name or Material Manganese ICP Standard
Show More Show Less