missing translation for 'onlineSavingsMsg'
Learn More

Manganese ICP-MS Standard, 1000 ppm Mn in 3% HNO3, Ricca Chemical

Catalog No. MSMN1KN100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSMN1KN100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSMN1KN100 Supplier Ricca Chemical Company Supplier No. MSMN1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6156-78-1 , 7697-37-2 , 7732-18-5
Molecular Formula C4H14MnO8
Molecular Weight (g/mol) 245.09
MDL Number MFCD00062552
InChI Key CESXSDZNZGSWSP-UHFFFAOYSA-L
PubChem CID 93021
IUPAC Name manganese(2+) diacetate tetrahydrate
SMILES O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Mn
CAS Min % 93.0, 4.21, 0.42
CAS Max % 97.0, 4.38, 0.43
Equivalence 1 mL = 1 mg Mn
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type ICP Single-element Standard Solution
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Manganese ICP-MS Standard
Show More Show Less