missing translation for 'onlineSavingsMsg'
Learn More

Manganese Gluconate, Dihydrate, USP, 98-102%, Spectrum™ Chemical

Catalog No. 18604192
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604192 50 kg Fiber Drum
18604189 500 g Poly Bottle
18604190 2.5 kg Poly Pail
18604191 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604192 Supplier Spectrum Chemical Mfg Cor Supplier No. M140450KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M1404, 6485-39-8, C12H22MnO14·2H2O

Spectrum™ Chemical Manganese Gluconate, Dihydrate, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6485-39-8

Specifications

CAS Max % 100%
Quantity 50 kg
Percent Purity 98 to 102%
Grade USP
Packaging Fiber Drum