missing translation for 'onlineSavingsMsg'
Learn More

Manganese Chloride Tetrahydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. M87500
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M87500 500 g Glass Bottle
M87100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M87500 Supplier Thermo Fisher Scientific Supplier No. M87 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
Synonym Manganese (II) Chloride
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Melting Point 58°C
Color Red
pH 5.5
Physical Form Solid
Quantity 500 g
Assay Percent Range 98 to 101%
Formula Weight 197.91
Percent Purity 98 to 101%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Density 2.01g/cm³
Chemical Name or Material Manganese Chloride Tetrahydrate
Show More Show Less