missing translation for 'onlineSavingsMsg'
Learn More

Manganese Carbonate, Powder, 43.5%, Spectrum™ Chemical

$418.00 - $26725.07

Chemical Identifiers

CAS 598-62-9
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18611478
View Documents
Spectrum Chemical Mfg Cor
M1100125GM
125 g Poly Bottle
Each for $418.00
Only null left
 
18611483
View Documents
Spectrum Chemical Mfg Cor
M1100500GM
500 g Poly Bottle
Each for $1,359.47
Only null left
 
18611481
View Documents
Spectrum Chemical Mfg Cor
M1100250LBBL
250 lb. Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18611480
View Documents
Spectrum Chemical Mfg Cor
M110025KG
2.5 kg Poly Bottle
Each for $4,514.87
Only null left
 
18611479
View Documents
Spectrum Chemical Mfg Cor
M110012KG
12 kg Poly Pail
Each for $5,507.33
Only null left
 
18611482
View Documents
Spectrum Chemical Mfg Cor
M110045KGBL
45 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

598-62-9

Specifications

598-62-9
125 g
Reagent
1
43.5%
Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.