missing translation for 'onlineSavingsMsg'
Learn More

Manganese Carbonate, Powder, 43.5%, Spectrum™ Chemical

Catalog No. 18611483
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
250 lb.
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611483 500 g Poly Bottle
18611478 125 g Poly Bottle
18611481 250 lb. Poly Bottle
18611480 2.5 kg Poly Bottle
18611479 12 kg Poly Bottle
18611482 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18611483 Supplier Spectrum Chemical Mfg Cor Supplier No. M1100500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 598-62-9 | 114.95 g/mol

Chemical Identifiers

CAS 598-62-9

Specifications

CAS Max % 1
Physical Form Powder, Needles or Crystals
Quantity 500 g
Molecular Weight (g/mol) 114.95
Percent Purity 43.5%
Grade Reagent
Packaging Poly Bottle
Chemical Name or Material Manganese Carbonate, Powder

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.