missing translation for 'onlineSavingsMsg'
Learn More

Manganese Carbonate, Powder, 43.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor M110045KGBL

 View more versions of this product

Catalog No. 18611482

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

598-62-9
45 kg
Reagent
1
43.5%
Fiber Drum

Chemical Identifiers

598-62-9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.