missing translation for 'onlineSavingsMsg'
Learn More

Manganese Carbonate, Powder, 43.5%, Spectrum™ Chemical

Catalog No. 18611478
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
250 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611478 125 g Poly Bottle
18611483 500 g Poly Bottle
18611481 250 lb. Fiber Drum
18611480 2.5 kg Poly Bottle
18611479 12 kg Poly Pail
18611482 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18611478 Supplier Spectrum Chemical Mfg Cor Supplier No. M1100125GM

Chemical Identifiers

CAS 598-62-9

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 43.5%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.