Learn More
Manganese AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Mn), LabChem™
Supplier: LabChem LC165501
Specifications
| Manganese AA Standard | |
| Poly Bottle | |
| 95.55,4,0.45 | |
| 1.05g/mL | |
| C4H14MnO8 | |
| C4H6MnO4·4H2O | |
| Colorless | |
| 102°C | |
| CESXSDZNZGSWSP-UHFFFAOYSA-L | |
| manganese(2+) diacetate tetrahydrate | |
| 500 mL | |
| 245.09 |
| 1000ppm (1mL = 1mg Mn) | |
| 6156-78-1 , 7697-37-2 , 7732-18-5 | |
| Passes Test | |
| Liquid | |
| 1000 ppm ±10 ppm (1mL = 1mg Mn) | |
| MFCD00062552 | |
| -7°C | |
| Soluble in water | |
| O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O | |
| 245.09 | |
| 93021 | |
| Certified |
Chemical Identifiers
| 6156-78-1 | |
| 245.09 | |
| CESXSDZNZGSWSP-UHFFFAOYSA-L | |
| manganese(2+) diacetate tetrahydrate |
| C4H14MnO8 | |
| MFCD00062552 | |
| 93021 | |
| O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature