missing translation for 'onlineSavingsMsg'
Learn More

Manganese AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Mn), LabChem™

Catalog No. LC165507
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC165507 125 mL Poly Bottle
LC165501 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC165507 Supplier LabChem Supplier No. LC165507

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6156-78-1 , 7697-37-2 , 7732-18-5
Molecular Formula C4H14MnO8
Molecular Weight (g/mol) 245.09
MDL Number MFCD00062552
InChI Key CESXSDZNZGSWSP-UHFFFAOYSA-L
PubChem CID 93021
IUPAC Name manganese(2+) diacetate tetrahydrate
SMILES O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O

Specifications

Packaging Poly Bottle
CAS Max % 95.55, 4, 0.45
Identification Passes Test
Density 1.05g/mL
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg Mn)
Linear Formula C4H6MnO4·4H2O
Color Colorless
Melting Point -7°C
Boiling Point 102°C
Solubility Soluble in water
Quantity 125 mL
Formula Weight 245.09
Grade Certified
Chemical Name or Material Manganese AA Standard, 1000ppm (1mL = 1mg Mn)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature