missing translation for 'onlineSavingsMsg'
Learn More

Mandelonitrile 97.0+%, TCI America™

Catalog No. M056625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M056625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M056625G Supplier TCI America Supplier No. M056625G
Only null left

Chemical Identifiers

CAS 532-28-5
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00004487
InChI Key NNICRUQPODTGRU-UHFFFAOYNA-N
Synonym mandelonitrile, benzaldehyde cyanohydrin, amygdalonitrile, phenylglycolonitrile, mandelic acid nitrile, glycolonitrile, phenyl, benzaldehydkyanhydrin, acetonitrile, hydroxyphenyl, nitril kyseliny mandlove, alpha-hydroxybenzeneacetonitrile
PubChem CID 10758
ChEBI CHEBI:16910
IUPAC Name 2-hydroxy-2-phenylacetonitrile
SMILES OC(C#N)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 102°C
Quantity 25 g
UN Number 3276
Formula Weight 133.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Mandelonitrile
EINECSNumber (3)-1823
RTECSNumber OO8400000
TSCA Yes