missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Mandelamide, 97%

Catalog No. AAL0467906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0467906 5 g
AAL0467903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0467906 Supplier Thermo Scientific Chemicals Supplier No. L0467906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4410-31-5 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4410-31-5
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00025495
InChI Key MAGPZHKLEZXLNU-UHFFFAOYSA-N
Synonym mandelamide, dl-mandelamide, o-hydroxy phenylacetamide, 2-hydroxy-2-phenyl-acetamide, benzeneacetamide, .alpha.-hydroxy, hydrate, -mandelamide, mandelic acid amide, +/--mandelamide
PubChem CID 73558
IUPAC Name 2-hydroxy-2-phenylacetamide
SMILES C1=CC=C(C=C1)C(C(=O)N)O

Specifications

Melting Point 132°C to 135°C
Quantity 5 g
Beilstein 2208677
Formula Weight 151.16
Percent Purity 97%
Chemical Name or Material (+/-)-Mandelamide
EINECSNumber 626-245-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only