missing translation for 'onlineSavingsMsg'
Learn More

Malonic Acid Reagent, For Ozone, LabChem™

Supplier:  LabChem LC165207

Catalog No. LC165207

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Malonic Acid Reagent
For Ozone
95,5
C3H4O4
malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
OFOBLEOULBTSOW-UHFFFAOYSA-N
propanedioic acid
867
104.06
Poly Bottle
Carbon monoxide; Carbon dioxide
Liquid
Colorless
141-82-2,7732-18-5
C3H4O4
1.05g/mL
Soluble in water
C(C(=O)O)C(=O)O
104.061
CHEBI:30794
Passes Test
1.05g/mL
125 mL

Chemical Identifiers

141-82-2
104.061
malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
CHEBI:30794
C(C(=O)O)C(=O)O
C3H4O4
OFOBLEOULBTSOW-UHFFFAOYSA-N
867
propanedioic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear eye protection.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions