accessibility menu, dialog, popup

UserName

OHOOHO

CAS RN 141-82-2

IUPAC Name: propanedioic acid
Synonyms: malonic acid propanediolic acid H2malo metahnedicarboxylic acid dicarboxylate dicarboxylic acid C3
Molecular Weight (g/mol): 104.06
Molecular Formula: C3H4O4
InChi Key: OFOBLEOULBTSOW-UHFFFAOYSA-N
SMILES: OC(=O)CC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)
Percent Purity 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results