missing translation for 'onlineSavingsMsg'
Learn More

Malic Acid, FCC, 99-100.5%, Spectrum™ Chemical

Catalog No. 18604150
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604150 500 g Poly Bottle
18604151 2.5 kg Poly Pail
18604152 12 kg Poly Pail
18604153 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604150 Supplier Spectrum Chemical Mfg Cor Supplier No. MA147500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

MA147, 617-48-1, C4H6O5

Spectrum™ Chemical Malic Acid, FCC is used as a source of extreme tartness. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 617-48-1
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.09
InChI Key BJEPYKJPYRNKOW-UHFFFAOYNA-N
IUPAC Name 2-hydroxybutanedioic acid
SMILES OC(CC(O)=O)C(O)=O

Specifications

CAS Max % 100%
Percent Purity 99 to 100.5%
Grade FCC
Quantity 500 g
Ignition Residue 0.1%
Optical Rotation -0.10° to +0.10°
Packaging Poly Bottle