missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maleic Hydrazide 99.0+%, TCI America™
Spécifications
Spécifications
| Color | White to Yellow |
| pH | 3.20 (50 g/L H2O soln, 25°C) |
| Quantity | 500 g |
| Formula Weight | 112.09 |
| Percent Purity | ≥99.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Maleic Hydrazide |
Sécurité et manipulation
| EINECSNumber | (5)-0973 |
| RTECSNumber | UR5950000 |
| TSCA | Yes |