missing translation for 'onlineSavingsMsg'
Learn More

Maleic Acid, BAKER™, J.T. Baker™

Catalog No. 02003279
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003279 500 g Poly Bottle
02003278 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02003279 Supplier Avantor Supplier No. P46007

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 110-16-7
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00063177
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
Synonym maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid
PubChem CID 444266
ChEBI CHEBI:18300
IUPAC Name (2E)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(O)=O

Specifications

Formula Weight 116.07
Quantity 500 g
Packaging Poly Bottle
Specific Gravity 1L = 1.59kg
Chemical Name or Material Maleic Acid