missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Malachite Green oxalate, pure, certified
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Absorption Ratio | 1.00 to 1.15 |
| Dye Content | 90% min. |
| Beilstein | 13, 745 |
| Merck Index | 15, 5763 |
| Infrared Spectrum | Authentic |
| Lambda Max | 614 to 620nm (in water) |
| Formula Weight | 927.03 |
| Grade | Pure |
| Packaging | Glass Bottle |
| Vapor Pressure | Vapor Pressure: Negligible |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: Toxic if swallowed. Suspected of damaging the unborn child. Causes serious eye damage. Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | Emergency Overview Toxic if swallowed. Causes eye burns. Risk of serious damage to eyes. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. May cause harm to the unborn |
RUO – Research Use Only