missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Streptavidin Positive Selection Beads

Catalog No. 501125939
Change view
Click to view available options
Quantity:
200 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125939 200 Tests
501125938 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Contains Sodium azide
Catalog No. 501125939 Supplier Invitrogen™ Supplier No. MSPB600374

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains Sodium azide

Designed for the magnetic separation of cells by positive selection. Desired cells are bound by biotinylated antibody and then streptavidin-coated magnetic beads. When placed in a magnetic field, desired cells are held in place and undesired cells can be separated by decanting.

  • Clone and concentration of biotinylated antibody for desired cell labeling, as well as concentration of MagniSort™ Streptavidin Positive Selection Beads must be determined empirically
  • Please refer to the protocol for details on how to optimize both the antibody and MagniSort™ Streptavidin Positive Selection Beads
  • The MagniSort™ Streptavidin Positive Selection Beads may also be used to deplete undesired cells using a single antibody
  • For depletion of cells using multiple antibodies, we recommend the MagniSort™ Streptavidin Negative Selection Beads (cat. MSNB-6002) and protocol

Specifications

Quantity 200 Tests
Form Liquid
Ligand Streptavidin
Product Type Streptavidin Conjugate (Fluorescent)
Target Molecule Biotin

For Research Use Only. Not for use in diagnostic procedures.