missing translation for 'onlineSavingsMsg'
Learn More

Magnesium, turnings, 99+%, Thermo Scientific Chemicals

Catalog No. AAL0812036
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0812036 500 g
AAL081200E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0812036 Supplier Thermo Scientific Chemicals Supplier No. L0812036

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Magnesium, turnings is used in solid state synthesis with Ca and Sn resulted in a new phase, Ca6.2Mg3.8Sn7, which has an unprecedented type of tin chain composed of square-planar tin units. It is a component in die-casting, to remove sulfur in the production of iron and steel, the production of titanium in the Kroll process. It is widely used for manufacturing of mobile phones, laptop and tablet computers, cameras, and other electronic components.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium, turnings is used in solid state synthesis with Ca and Sn resulted in a new phase, Ca6.2Mg3.8Sn7, which has an unprecedented type of tin chain composed of square-planar tin units. It is a component in die-casting, to remove sulfur in the production of iron and steel, the production of titanium in the Kroll process. It is widely used for manufacturing of mobile phones, laptop and tablet computers, cameras, and other electronic components.

Solubility
Reacts with water.

Notes
Store away from air, moisture and oxidizing agents. Incompatible with acids, air, nitrogen and moisture.

Chemical Identifiers

CAS 7439-95-4
Molecular Formula Mg
Molecular Weight (g/mol) 24.30
MDL Number MFCD00085308
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
Synonym magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs
PubChem CID 5462224
ChEBI CHEBI:25107
IUPAC Name magnesium
SMILES [Mg]

Specifications

Melting Point 650°C
Boiling Point 1091°CC
Density 1.738
Quantity 500 g
Odor Odorless
Sensitivity Moisture sensitive
UN Number UN1869
Beilstein 4948473
Solubility Information Reacts with water.
Formula Weight 24.305
Percent Purity ≥99%
Chemical Name or Material Magnesium, turnings
Show More Show Less
Hazard Category H228-H252-H261
Hazard Statement GHS H Statement
H228-H251-H261
Flammable solid.
Self-heating: may catch fire.
In contact with water releases flammable gas.
Precautionary Statement P210-P223-P231+P232-P235+P410-P240-P241-P280-P335+P334-P370+P378s-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: MAGNESIUM
EINECSNumber 231-104-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only